2-Amino-N-benzyl-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

053396-500MG 053396-2.5G
101903-470EA 364.62 USD
101903-470 101903-472
2-Amino-N-benzyl-3-methylbutanamide hydrochloride
2-Amino-N-benzyl-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O
MDL Number: MFCD13562757
CAS Number: 1236262-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR