3-Amino-N-benzylpropanamide hydrochloride
Supplier: Matrix Scientific
053395-500MG
053395-2.5G
101903-466EA
364.62
USD
101903-466
101903-468
3-Amino-N-benzylpropanamide hydrochloride
3-Amino-N-benzylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O |
MDL Number:
MFCD13562756 CAS Number: 202819-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...