N-Benzyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

053394-500MG 053394-2.5G
101903-462EA 364.62 USD
101903-462 101903-464
N-Benzyl-2-(methylamino)acetamide hydrochloride
N-Benzyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD13562755
CAS Number: 860217-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR