2-Amino-N-benzylpropanamide hydrochloride

Supplier: Matrix Scientific

053393-500MG 053393-2.5G
101903-458EA 149.23 USD
101903-458 101903-460
2-Amino-N-benzylpropanamide hydrochloride
2-Amino-N-benzylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD13195941
CAS Number: 1214706-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR