2-Amino-N-benzylacetamide hydrochloride

Supplier: Matrix Scientific

053392-500MG 053392-2.5G
101903-454EA 384.62 USD
101903-454 101903-456
2-Amino-N-benzylacetamide hydrochloride
2-Amino-N-benzylacetamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₃ClN₂O
MDL Number: MFCD09909316
CAS Number: 20432-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR