N-Benzyl-5-bromo-3-methyl-2-pyridinamine
Supplier: Matrix Scientific
053389-500MG
053389-2.5G
101903-442EA
364.62
USD
101903-442
101903-444
N-Benzyl-5-bromo-3-methyl-2-pyridinamine
N-Benzyl-5-bromo-3-methyl-2-pyridinamine
Hazard: Irritant
Formula:
C₁₃H₁₃BrN₂ |
MDL Number:
MFCD13562754 CAS Number: 1219976-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...