N-Benzyl-2-chloronicotinamide
Supplier: Matrix Scientific
Synonyms:
N3-Benzyl-2-chloronicotinamide
053388-500MG
053388-2.5G
101903-438EA
364.62
USD
101903-438
101903-440
N-Benzyl-2-chloronicotinamide
N-Benzyl-2-chloronicotinamide
Hazard: Irritant
Formula:
C₁₃H₁₁ClN₂O MW: 246.7 g/mol |
MDL Number:
MFCD00831758 CAS Number: 65423-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...