N2-Benzyl-2,3-pyridinediamine

Supplier: Matrix Scientific

053387-500MG 053387-2.5G
101903-434EA 166.15 USD
101903-434 101903-436
N2-Benzyl-2,3-pyridinediamine
N2-Benzyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₂H₁₃N₃
MDL Number: MFCD09757497
CAS Number: 32282-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR