N-Benzyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine
Supplier: Matrix Scientific
053384-500MG
053384-2.5G
101903-422EA
364.62
USD
101903-422
101903-424
N-Benzyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine
N-Benzyl-3-chloro-5-(trifluoromethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₃H₁₀ClF₃N₂ |
MDL Number:
MFCD12618435 CAS Number: 1041602-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...