N-Benzyl-6-chloro-4-pyrimidinamine
Supplier: Matrix Scientific
053383-500MG
053383-2.5G
101903-418EA
100
USD
101903-418
101903-420
N-Benzyl-6-chloro-4-pyrimidinamine
N-Benzyl-6-chloro-4-pyrimidinamine
Hazard: Irritant
Formula:
C₁₁H₁₀ClN₃ |
MDL Number:
MFCD00679879 CAS Number: 61667-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...