4-(Benzylamino)-3-nitrobenzoic acid

Supplier: Matrix Scientific

053381-500MG 053381-2.5G
101903-410EA 364.62 USD
101903-410 101903-412
4-(Benzylamino)-3-nitrobenzoic acid
4-(Benzylamino)-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₄
MDL Number: MFCD01156575
CAS Number: 68502-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR