Ethyl-3-amino-4-(benzylamino)benzoate
Supplier: Matrix Scientific
053380-500MG
053380-2.5G
101903-406EA
364.62
USD
101903-406
101903-408
Ethyl-3-amino-4-(benzylamino)benzoate
Ethyl-3-amino-4-(benzylamino)benzoate
Hazard: Irritant
Formula:
C₁₆H₁₈N₂O₂ |
MDL Number:
MFCD13562752 CAS Number: 848819-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...