N-(4-Methylphenyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053377-500MG 053377-2.5G
101903-394EA 364.62 USD
101903-394 101903-396
N-(4-Methylphenyl)-4-piperidinecarboxamide hydrochloride
N-(4-Methylphenyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD13562750
CAS Number: 1220035-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR