N-(4-Methylphenyl)-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053376-500MG
053376-2.5G
101903-390EA
364.62
USD
101903-390
101903-392
N-(4-Methylphenyl)-3-piperidinecarboxamide hydrochloride
N-(4-Methylphenyl)-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O |
MDL Number:
MFCD13562749 CAS Number: 599184-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...