N-(4-Methylphenyl)-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053374-500MG 053374-2.5G
101903-382EA 364.62 USD
101903-382 101903-384
N-(4-Methylphenyl)-2-pyrrolidinecarboxamide hydrochloride
N-(4-Methylphenyl)-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD13562747
CAS Number: 1236257-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR