2-Amino-N-(4-methylphenyl)-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

053373-500MG 053373-2.5G
101903-378EA 364.62 USD
101903-378 101903-380
2-Amino-N-(4-methylphenyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(4-methylphenyl)-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₉ClN₂O
MDL Number: MFCD13562746
CAS Number: 1236256-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR