2-Amino-2-methyl-N-(4-methylphenyl)propanamide hydrochloride

Supplier: Matrix Scientific

053372-500MG 053372-2.5G
101903-374EA 364.62 USD
101903-374 101903-376
2-Amino-2-methyl-N-(4-methylphenyl)propanamide hydrochloride
2-Amino-2-methyl-N-(4-methylphenyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O
MDL Number: MFCD13562745
CAS Number: 1220030-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR