3-Amino-N-(4-methylphenyl)propanamide hydrochloride
Supplier: Matrix Scientific
053370-500MG
053370-2.5G
101903-366EA
364.62
USD
101903-366
101903-368
3-Amino-N-(4-methylphenyl)propanamide hydrochloride
3-Amino-N-(4-methylphenyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O |
MDL Number:
MFCD13194007 CAS Number: 515131-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...