2-(Methylamino)-N-(4-methylphenyl)acetamide hydrochloride

Supplier: Matrix Scientific

053369-500MG 053369-2.5G
101903-362EA 364.62 USD
101903-362 101903-364
2-(Methylamino)-N-(4-methylphenyl)acetamide hydrochloride
2-(Methylamino)-N-(4-methylphenyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD08445327
CAS Number: 64642-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR