2-Amino-N-(4-methylphenyl)propanamide hydrochloride
Supplier: Matrix Scientific
053368-500MG
053368-2.5G
101903-358EA
364.62
USD
101903-358
101903-360
2-Amino-N-(4-methylphenyl)propanamide hydrochloride
2-Amino-N-(4-methylphenyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O |
MDL Number:
MFCD13562743 CAS Number: 1236259-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...