5-Bromo-3-methyl-N-(4-methylphenyl)-2-pyridinamine

Supplier: Matrix Scientific

053364-500MG 053364-2.5G
101903-342EA 364.62 USD
101903-342 101903-344
5-Bromo-3-methyl-N-(4-methylphenyl)-2-pyridinamine
5-Bromo-3-methyl-N-(4-methylphenyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₃BrN₂
MDL Number: MFCD13562739
CAS Number: 1220020-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR