2-Chloro-N-(4-methylphenyl)nicotinamide

Supplier: Matrix Scientific

053363-500MG 053363-2.5G
101903-338EA 364.62 USD
101903-338 101903-340
2-Chloro-N-(4-methylphenyl)nicotinamide
2-Chloro-N-(4-methylphenyl)nicotinamide

Hazard: Irritant

Formula: C₁₃H₁₁ClN₂O
MDL Number: MFCD03059168
CAS Number: 56149-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR