2-(4-Toluidino)nicotinic acid

Supplier: Matrix Scientific

053361-500MG 053361-2.5G
101903-330EA 384.62 USD
101903-330 101903-332
2-(4-Toluidino)nicotinic acid
2-(4-Toluidino)nicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₂
MDL Number: MFCD06016518
CAS Number: 60645-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR