2-(4-Toluidino)isonicotinic acid

Supplier: Matrix Scientific

053360-500MG 053360-2.5G
101903-326EA 364.62 USD
101903-326 101903-328
2-(4-Toluidino)isonicotinic acid
2-(4-Toluidino)isonicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₂
MW: 228.25 g/mol
MDL Number: MFCD11132177
CAS Number: 1019452-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR