6-Chloro-N-(4-methylphenyl)-2-pyrazinamine

Supplier: Matrix Scientific

053359-500MG 053359-2.5G
101903-322EA 149.23 USD
101903-322 101903-324
6-Chloro-N-(4-methylphenyl)-2-pyrazinamine
6-Chloro-N-(4-methylphenyl)-2-pyrazinamine

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃
MDL Number: MFCD10613403
CAS Number: 1030187-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR