6-Chloro-N-(4-methylphenyl)-4-pyrimidinamine
Supplier: Matrix Scientific
053357-500MG
053357-2.5G
101903-314EA
364.62
USD
101903-314
101903-316
6-Chloro-N-(4-methylphenyl)-4-pyrimidinamine
6-Chloro-N-(4-methylphenyl)-4-pyrimidinamine
Hazard: Irritant
Formula:
C₁₁H₁₀ClN₃ |
MDL Number:
MFCD13562737 CAS Number: 1219976-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...