5-Bromo-4-methyl-N-(4-methylphenyl)-2-pyridinamine

Supplier: Matrix Scientific

053356-500MG 053356-2.5G
101903-310EA 364.62 USD
101903-310 101903-312
5-Bromo-4-methyl-N-(4-methylphenyl)-2-pyridinamine
5-Bromo-4-methyl-N-(4-methylphenyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₃BrN₂
MDL Number: MFCD13562736
CAS Number: 1220017-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR