3-Nitro-4-(4-toluidino)benzoic acid

Supplier: Matrix Scientific

053355-500MG 053355-2.5G
101903-306EA 364.62 USD
101903-306 101903-308
3-Nitro-4-(4-toluidino)benzoic acid
3-Nitro-4-(4-toluidino)benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₄
MDL Number: MFCD01113675
CAS Number: 148304-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR