Ethyl-3-amino-4-(4-toluidino)benzoate
Supplier: Matrix Scientific
053354-500MG
053354-2.5G
101903-302EA
364.62
USD
101903-302
101903-304
Ethyl-3-amino-4-(4-toluidino)benzoate
Ethyl-3-amino-4-(4-toluidino)benzoate
Hazard: Irritant
Formula:
C₁₆H₁₈N₂O₂ |
MDL Number:
MFCD13562735 CAS Number: 1220020-00-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...