N-(3-Methylphenyl)-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053351-500MG 053351-2.5G
101903-290EA 364.62 USD
101903-290 101903-292
N-(3-Methylphenyl)-4-piperidinecarboxamide hydrochloride
N-(3-Methylphenyl)-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD13562733
CAS Number: 1220036-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR