N-(3-Methylphenyl)-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053350-500MG
053350-2.5G
101903-286EA
364.62
USD
101903-286
101903-288
N-(3-Methylphenyl)-3-piperidinecarboxamide hydrochloride
N-(3-Methylphenyl)-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O |
MDL Number:
MFCD13562732 CAS Number: 1220020-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...