N-(3-Methylphenyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053349-500MG 053349-2.5G
101903-282EA 364.62 USD
101903-282 101903-284
N-(3-Methylphenyl)-2-piperidinecarboxamide hydrochloride
N-(3-Methylphenyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MW: 254.76 g/mol
MDL Number: MFCD13562731
CAS Number: 1236265-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR