N-(3-Methylphenyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053348-500MG
053348-2.5G
101903-278EA
364.62
USD
101903-278
101903-280
N-(3-Methylphenyl)-2-pyrrolidinecarboxamide hydrochloride
N-(3-Methylphenyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇ClN₂O |
MDL Number:
MFCD13562730 CAS Number: 1236259-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...