2-Amino-N-(3-methylphenyl)-3-phenylpropanamide hydrochloride

Supplier: Matrix Scientific

053347-500MG 053347-2.5G
101903-274EA 364.62 USD
101903-274 101903-276
2-Amino-N-(3-methylphenyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(3-methylphenyl)-3-phenylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₉ClN₂O
MDL Number: MFCD13562729
CAS Number: 1236262-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR