3-Amino-N-(3-methylphenyl)propanamide hydrochloride

Supplier: Matrix Scientific

053344-500MG 053344-2.5G
101903-262EA 364.62 USD
101903-262 101903-264
3-Amino-N-(3-methylphenyl)propanamide hydrochloride
3-Amino-N-(3-methylphenyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD13194009
CAS Number: 938515-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR