2-(Methylamino)-N-(3-methylphenyl)acetamide hydrochloride
Supplier: Matrix Scientific
053343-500MG
053343-2.5G
101903-258EA
364.62
USD
101903-258
101903-260
2-(Methylamino)-N-(3-methylphenyl)acetamide hydrochloride
2-(Methylamino)-N-(3-methylphenyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O |
MDL Number:
MFCD13562726 CAS Number: 1220038-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...