2-Amino-N-(3-methylphenyl)propanamide hydrochloride
Supplier: Matrix Scientific
053342-500MG
053342-2.5G
101903-254EA
364.62
USD
101903-254
101903-256
2-Amino-N-(3-methylphenyl)propanamide hydrochloride
2-Amino-N-(3-methylphenyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₅ClN₂O |
MDL Number:
MFCD13562725 CAS Number: 1236261-20-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...