2-Amino-N-(3-methylphenyl)acetamide hydrochloride
Supplier: Matrix Scientific
053341-500MG
053341-2.5G
101903-250EA
364.62
USD
101903-250
101903-252
2-Amino-N-(3-methylphenyl)acetamide hydrochloride
2-Amino-N-(3-methylphenyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃ClN₂O |
MDL Number:
MFCD13562724 CAS Number: 1219964-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...