N-(5-Bromo-2-pyridinyl)-N-(3-methylphenyl)amine
Supplier: Matrix Scientific
053339-500MG
053339-2.5G
101903-242EA
364.62
USD
101903-242
101903-244
N-(5-Bromo-2-pyridinyl)-N-(3-methylphenyl)amine
N-(5-Bromo-2-pyridinyl)-N-(3-methylphenyl)amine
Hazard: Irritant
Formula:
C₁₂H₁₁BrN₂ |
MDL Number:
MFCD13562722 CAS Number: 1219964-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...