2-Chloro-N-(3-methylphenyl)nicotinamide
Supplier: Matrix Scientific
053337-500MG
053337-2.5G
101903-234EA
364.62
USD
101903-234
101903-236
2-Chloro-N-(3-methylphenyl)nicotinamide
2-Chloro-N-(3-methylphenyl)nicotinamide
Hazard: Irritant
Formula:
C₁₃H₁₁ClN₂O |
MDL Number:
MFCD03381804 CAS Number: 57841-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...