N2-(3-Methylphenyl)-2,3-pyridinediamine

Supplier: Matrix Scientific

053336-500MG 053336-2.5G
101903-230EA 364.62 USD
101903-230 101903-232
N2-(3-Methylphenyl)-2,3-pyridinediamine
N2-(3-Methylphenyl)-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₂H₁₃N₃
MDL Number: MFCD11202103
CAS Number: 176032-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR