2-(3-Toluidino)nicotinic acid
Supplier: Matrix Scientific
053335-500MG
053335-2.5G
101903-226EA
364.62
USD
101903-226
101903-228
2-(3-Toluidino)nicotinic acid
2-(3-Toluidino)nicotinic acid
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₂ |
MDL Number:
MFCD03114307 CAS Number: 57978-46-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...