2-(3-Toluidino)isonicotinic acid
Supplier: Matrix Scientific
053334-500MG
053334-2.5G
101903-222EA
364.62
USD
101903-222
101903-224
2-(3-Toluidino)isonicotinic acid
2-(3-Toluidino)isonicotinic acid
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₂ |
MDL Number:
MFCD11132194 CAS Number: 1019387-47-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...