6-Chloro-N-(3-methylphenyl)-2-pyrazinamine

Supplier: Matrix Scientific

053333-500MG 053333-2.5G
101903-218EA 364.62 USD
101903-218 101903-220
6-Chloro-N-(3-methylphenyl)-2-pyrazinamine
6-Chloro-N-(3-methylphenyl)-2-pyrazinamine

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃
MDL Number: MFCD13562720
CAS Number: 1220030-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR