6-Chloro-N-(3-methylphenyl)-4-pyrimidinamine

Supplier: Matrix Scientific

053331-500MG 053331-2.5G
101903-210EA 364.62 USD
101903-210 101903-212
6-Chloro-N-(3-methylphenyl)-4-pyrimidinamine
6-Chloro-N-(3-methylphenyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃
MDL Number: MFCD13562718
CAS Number: 169286-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR