3-Nitro-4-(3-toluidino)benzoic acid

Supplier: Matrix Scientific

053329-500MG 053329-2.5G
101903-202EA 364.62 USD
101903-202 101903-204
3-Nitro-4-(3-toluidino)benzoic acid
3-Nitro-4-(3-toluidino)benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₄
MDL Number: MFCD02939944
CAS Number: 452088-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR