Ethyl-3-amino-4-(3-toluidino)benzoate
Supplier: Matrix Scientific
053328-500MG
053328-2.5G
101903-198EA
364.62
USD
101903-198
101903-200
Ethyl-3-amino-4-(3-toluidino)benzoate
Ethyl-3-amino-4-(3-toluidino)benzoate
Hazard: Irritant
Formula:
C₁₆H₁₈N₂O₂ |
MDL Number:
MFCD13562716 CAS Number: 1220030-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...