3-Amino-4-chloro-N-(2-methylphenyl)benzenesulfonamide

Supplier: Matrix Scientific

053327-500MG 053327-2.5G
101903-194EA 364.62 USD
101903-194 101903-196
3-Amino-4-chloro-N-(2-methylphenyl)benzenesulfonamide
3-Amino-4-chloro-N-(2-methylphenyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₃H₁₃ClN₂O₂S
MW: 296.78 g/mol
MDL Number: MFCD12442165
CAS Number: 1041512-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR