N-(2-Methylphenyl)-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053324-500MG
053324-2.5G
101903-182EA
364.62
USD
101903-182
101903-184
N-(2-Methylphenyl)-3-piperidinecarboxamide hydrochloride
N-(2-Methylphenyl)-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O |
MDL Number:
MFCD13562713 CAS Number: 1220019-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...