N-(2-Methylphenyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053322-500MG
053322-2.5G
101903-174EA
364.62
USD
101903-174
101903-176
N-(2-Methylphenyl)-2-pyrrolidinecarboxamide hydrochloride
N-(2-Methylphenyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇ClN₂O |
MDL Number:
MFCD13562711 CAS Number: 1219100-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...